Skip to Content
Merck
All Photos(1)

Key Documents

218725

Sigma-Aldrich

Caspase-5, Human, Recombinant, E. coli

Caspase-5, Human, Recombinant, is expressed in E. coli. It is an inflammatory caspase.

Synonym(s):

ICErel-III, TY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

Assay

≥80% (SDS-PAGE)

form

liquid

specific activity

≥100,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

M.W. 10,000 and 22,000 (heterodimer).
Recombinant, human caspase-5 expressed in E. coli. A member of the caspase-1 subfamily of cysteine proteases involved in Fas-mediated apoptosis. Caspase-5 cleavage site is reported to be Asp311 and Asp330. May also cleave at Asp121. Suitable for the study of enzyme regulation and kinetics, target substrate cleavage, and inhibitor screening. MW: 10000 and 22000.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1.0 pmol substrate Ac-LEHD-pNA (Cat. No. 218805) per min at 25°C, pH 7.4.

Physical form

In 100 mM NaCl, 50 mM HEPES, 10 mM DTT, 1 mM EDTA, 10% glycerol, 0.5% CHAPS, pH 7.4.

Reconstitution

After initial defrost, aliquot and freeze at -70°C.

Other Notes

Schwartz, S. et al. 1999. Cancer Res. 59, 2995.
Fucheu, C., et al. 1996. Eur. J. Biochem. 236, 207.
Munday, N.A., et al. 1995. J. Biol. Chem. 270, 15870.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service