Skip to Content
Merck
All Photos(1)

Key Documents

2145-OP

Millipore

Ammonium Acetate

OmniPur® Grade

Synonym(s):

OmniPur® Ammonium Acetate, Acetic Acid, Ammonium Salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
CHF 116.00

CHF 116.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
1 KG
CHF 116.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

CHF 116.00


Estimated to ship on17 April 2025


Request a Bulk Order

grade

reagent

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

wet ice

storage temp.

10-30°C

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

General description

Molecular biology grade.

Specifications:
A (280 nm, 1 M, water): ≤0.01AU

A (354 nm, 1 M, water): ≤0.01AU
Assay: ≥99%

Chloride (Cl): ≤0.0005%
DNase: None detected
Heavy metals (as Pb): ≤0.0005%
Residue after ignition: ≤0.01%
Insoluble matter: ≤0.005%
Iron (Fe): ≤0.0005%
Nitrate (NO3): ≤0.001%
pH (5%, water, 25°C): 6.7-7.3
Potassium (K): ≤0.005%
Protease: None detected
RNase: None detected
Sodium (Na): ≤0.005%
Sulfate (SO4): ≤0.001%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • A green analytical method for the simultaneous determination of 17 perfluoroalkyl substances (PFAS) in human serum and semen by ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS).: Ammonium acetate is used as a key component in the mobile phase for UPLC-MS/MS, enhancing the detection and analysis of PFAS, underscoring its importance in environmental and health-related analytical chemistry (Di Giorgi A et al., 2024).

  • Innovation of 6-sulfonamide-2H-chromene derivatives as antidiabetic agents targeting a-amylase, a-glycosidase, and PPAR-y inhibitors with in silico molecular docking simulation.: Demonstrates the role of ammonium acetate in the synthesis of complex molecules with potential therapeutic applications, highlighting its use in medicinal chemistry for drug design and discovery (Thabet HK et al., 2024).

  • Regulation of Tetramethylpyrazine Formation by the Phenolics-Fenton Coupled Redox Cycling System.: Discusses the application of ammonium acetate in food chemistry, specifically in enhancing chemical reactions that influence flavor compound synthesis, which is critical for food science research (Xu H et al., 2024).

  • Simultaneous determination of unecritinib (TQ-B3101) and its active metabolite crizotinib in rat plasma by LC-MS/MS: An application to pharmacokinetic studies.: Utilizes ammonium acetate in the sample preparation phase for LC-MS/MS, facilitating the pharmacokinetic analysis of cancer treatments, thus contributing to oncological research and therapeutic monitoring (Wang H et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service