Skip to Content
Merck
All Photos(1)

Documents

207003

Sigma-Aldrich

Calcineurin Inhibitor VIII, CN585

The Calcineurin Inhibitor VIII, CN585, also referenced under CAS 1213234-31-1, controls the biological activity of Calcineurin. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Calcineurin Inhibitor VIII, CN585, PP2B Inhibitor VIII, Ca 2+/Calmodulin-dependent Protein Phosphatase Inhibitor, 6-(3,4-Dichlorophenyl)-4-(N,N-dimethylaminoethylthio)-2-phenyl-pyrimidine, PP2B Inhibitor VIII, Ca2+/Calmodulin-dependent Protein Phosphatase Inhibitor, 6-(3,4-Dichlorophenyl)-4-(N,N-dimethylaminoethylthio)-2-phenyl-pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19Cl2N3S
CAS Number:
Molecular Weight:
404.36
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable pyrimidine compound that is shown to inhibit the calcineurin (PP2B)-catalyzed dephosphorylation of RII phosphopeptide (Cat. No. 207008), but not pNPP (Cat. Nos. 4876, 487600, 487655, 487663, 487666), in a reversible, calmodulin-independent, and substrate-noncompetitive manner (IC50 = 3.5 µM; Ki = 3.8 µM), while exhibiting little activity against PP1, PP2A, or PP2C even at concentrations as high as 100 µM. Unlike CsA (Cat. No. 239835) and FK506 (Cat. No. 342500), CN585 does not require Cyp A (Cat. No. 239777) and FKBP family immunophilins for PP2B inhibition, nor does CN585 inhibit the peptidyl prolyl cis/trans isomerase (PPIase) activity of Cyp18, FKBP12 (Cat. No. 325902), and Pin1, although binding studies indicate that CN585 and Cyp18/CsA target a common or overlapping binding sites on the catalytic subunit of PP2B. CN585, at 30 µM, is shown to effectively block NFAT nuclear translocation in HeLa cells, NFAT transcription activity in Jurkat cells (IC50 = 10 µM), as well as IL-2 production in PBMC (IC50 = 13 µM) upon PMA/ionomycin stimulation.
A cell-permeable pyrimidine compound that is shown to inhibit the calcineurin (PP2B)-catalyzed dephosphorylation of RII phosphopeptide (Cat. No. 207008), but not pNPP (Cat. Nos. 4876, 487600, 487655, 487663, 487666), in a reversible, calmodulin-independent, and substrate-noncompetitive manner (IC50 = 3.5 µM; Ki = 3.8 µM), while exhibiting little activity against PP1, PP2A, or PP2C even at concentrations as high as 100 µM. Unlike CsA (Cat. No. 239835) and FK506 (Cat. No. 342500), CN585 does not require Cyp A (Cat. No. 239777) and FKBP family immunophilins for PP2B inhibition, nor does CN585 inhibit the peptidyl prolyl cis/trans isomerase (PPIase) activity of Cyp18, FKBP12 (Cat. No. 325902), and Pin1, although binding studies indicate that CN585 and Cyp18/CsA target a common or overlapping binding sites on the catalytic subunit of PP2B. CN585, at 30 µM, is shown to effectively block NFAT nuclear translocation in HeLa cells, NFAT transcription activity in Jurkat cells (IC50 = 10 µM), as well as IL-2 production in PBMC (IC50 = 13 µM) upon PMA/ionomycin stimulation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Erdmann, F., et al. 2010. J. Biol. Chem.285, 1888.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Masaki et al.
Scientific reports, 13(1), 13116-13116 (2023-08-13)
c-Myc, a transcription factor, induces cell proliferation and is often aberrantly or highly expressed in cancers. However, molecular mechanisms underlying this aberrantly high expression remain unclear. Here, we found that intracellular Ca2+ concentration regulates c-Myc oncoprotein stability. We identified that
Haruki Tomiyasu et al.
The Journal of biological chemistry, 300(4), 107209-107209 (2024-03-23)
FOXO1 is a transcription factor and potential tumor suppressor that is negatively regulated downstream of PI3K-PKB/AKT signaling. Paradoxically, FOXO also promotes tumor growth, but the detailed mechanisms behind this role of FOXO are not fully understood. In this study, we
Ilse Delint-Ramirez et al.
Molecular cell, 82(20), 3794-3809 (2022-10-08)
Neuronal activity induces topoisomerase IIβ (Top2B) to generate DNA double-strand breaks (DSBs) within the promoters of neuronal early response genes (ERGs) and facilitate their transcription, and yet, the mechanisms that control Top2B-mediated DSB formation are unknown. Here, we report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service