Skip to Content
Merck
All Photos(1)

Key Documents

205546

Sigma-Aldrich

Caffeic Acid

A natural dietary compound reported to have anti-carcinogenic and anti-inflammatory properties.

Synonym(s):

Caffeic Acid, 3,4-Dihydroxycinnamic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 134.00

CHF 134.00


Estimated to ship on28 May 2025



Select a Size

Change View
500 MG
CHF 134.00

About This Item

Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

CHF 134.00


Estimated to ship on28 May 2025


Quality Level

description

Merck USA index - 14, 1635

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

tan

solubility

DMSO: 200 mg/mL
ethanol: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Oc1c(ccc(c1)\C=C/C(=O)O)O

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2-

InChI key

QAIPRVGONGVQAS-RQOWECAXSA-N

General description

A cell-permeable natural dietary compound reported to have anti-carcinogenic and anti-inflammatory properties. A selective, non-competitive inhibitor of 5-lipoxygenase (ID50 = 3.7 µM). Inhibits Cu2+-induced LDL oxidation in the initiation phase but acts as a prooxidant in the propagation phase. An effective irreversible inhibitor of glutathione S-transferases and a non-competitive inhibitor of xanthine oxidase.
A natural dietary compound reported to have anti-carcinogenic and anti-inflammatory properties. A cell-permeable, selective, non-competitive inhibitor of 5-lipoxygenase (ID50 = 3.7 µM). Inhibits Cu2+-induced LDL oxidation in the initiation phase but acts as a prooxidant in the propagation phase. An irreversible inhibitor of glutathione S-transferases and a non-competitive inhibitor of xanthine oxidase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
5-lipoxygenase
Product does not compete with ATP.
Reversible: no
Target IC50: 3.7 µM against 5-lipoxygenase

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yamanaka, N., et al. 1997. FEBS Lett.405, 186.
Chan, W.S., et al. 1995. Anticancer Res.15, 703.
Ploemen, J.H., et al. 1993. Food Chem. Toxicol.31, 475.
Koshihara, Y., et al. 1984. Biochim. Biophys. Acta792, 92.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengzheng Xiao et al.
Molecular medicine reports, 23(5) (2021-03-03)
Recent studies have reported that gene amplified in squamous cell carcinoma 1 (GASC1) is involved in the progression of several types of cancer. However, whether GASC1 promotes glioma progression remains unknown. Therefore, the present study aimed to investigate the effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service