Skip to Content
Merck
All Photos(1)

Key Documents

196418

Millipore

BAPTA, Tetrasodium Salt

Ca2+ chelator exhibiting a 105-fold greater affinity for Ca2+ .

Synonym(s):

BAPTA, Tetrasodium Salt, 1,2- bis( o-Aminophenoxy)ethane-N,N,Nʹ,Nʹ-tetraacetic Acid, 4Na, 1,2-bis(o-Aminophenoxy)ethane-N,N,Nʹ,Nʹ-tetraacetic Acid, 4Na

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C22H20N2O10 · 4Na
CAS Number:
Molecular Weight:
564.36
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

form

solid

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].N(CC(=O)[O-])(CC(=O)[O-])c1c(cccc1)OCCOc2c(cccc2)N(CC(=O)[O-])CC(=O)[O-]

InChI

1S/C22H24N2O10.4Na/c25-19(26)11-23(12-20(27)28)15-5-1-3-7-17(15)33-9-10-34-18-8-4-2-6-16(18)24(13-21(29)30)14-22(31)32;;;;/h1-8H,9-14H2,(H,25,26)(H,27,28)(H,29,30)(H,31,32);;;;/q;4*+1/p-4

InChI key

ZWSMLJACYSGFKD-UHFFFAOYSA-J

General description

Ca2+-chelator exhibiting a 105-fold greater affinity for Ca2+ (110 nM) than for Mg2+. After BAPTA complexes with Ca2+, the UV spectrum shifts from 254 to 279 nm at pH >6.0. Useful for spectrophotometric monitoring of extracellular Ca2+ levels, especially transient phenomena, because of its insensitivity to pH. Has been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. In conjunction with certain formazan dyes, BAPTA can be used in a colorimetric assay for Mg2+. Has neuroprotective effects in vitro against excitotoxic damage and spontaneous recurrent seizures, such as epilepsy.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot, purge with inert gas and freeze (-20°C). To increase stability in solution, store at a high pH (>10). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

DeLorenzo, R.J., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 14482.
Li, L., et al. 1995. J. Cell Physiol. 163, 105.
Tymianski, M., et al. 1994. J. Neurophysiol. 72, 1973.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service