Skip to Content
Merck
All Photos(1)

Documents

1.93010

Supelco

Ascorbic acid

for analysis EMPARTA® ACS

Synonym(s):

L-Ascorbic acid, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
MDL number:
UNSPSC Code:
12352205

grade

ACS reagent

mp

190-194 °C (dec.)

storage temp.

room temp

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93010.0121: 100 g, Minimum order quantity: 20
1.93010.0521: 500 g, Minimum order quantity: 20
1.93010.5021: 5 kg, Minimum order quantity: 4
1.93010.9026: 25 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service