189485
Autocamtide-2 Related Inhibitory Peptide II, Cell-permeable
The Autocamtide-2 Related Inhibitory Peptide II, Cell-permeable controls the biological activity of calmodulin-dependent protein kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Autocamtide-2 Related Inhibitory Peptide II, Cell-permeable, Ant-A3K/V10F-AIP, Ant-AIP-II, Ac-RQIKIWFQNRRMKWKKKKKLRRQEAFDAL-OH
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
white
solubility
5% acetic acid: 1 mg/mL
water: 1 mg/mL
DMSO: 5 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
Biochem/physiol Actions
cam Kinase-2
Packaging
Warning
Sequence
Physical form
Reconstitution
Other Notes
Ishida, A., et al. 1998. FEBS Lett.427, 115.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service