18-152
Streptavidin, HRP conjugate
1 mg Streptavidin, HRP conjugate for ELISA, Western Blotting, ICC & IHC.
Synonym(s):
detection reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Upstate®
technique(s)
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
shipped in
wet ice
Application
Streptavidin, horseradish peroxidase (HRP) conjugate has been used in:
- cell-based enzyme linked immunosorbent assay (ELISA)
- sandwich ELISA
- microneutralization assay
Biochem/physiol Actions
Streptavidin antibody, covalently bound to horseradish peroxidase (HRP) is used in enzyme-linked detection methods. The conjugated enzyme catalyzes the breakdown of certain substrates, leading to signal generation.
Physical form
0.05M HEPES, 0.1M Na2SO4, pH6.8, preservative.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service