Skip to Content
Merck
All Photos(1)

Key Documents

1.59712

Supelco

Methoxyphenylacetic acid

Reag. Ph Eur

Synonym(s):

Methoxyphenylacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CHF 258.00

CHF 258.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 G
CHF 258.00

About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
230-300-9
NACRES:
NA.21

CHF 258.00


Estimated to ship on01 June 2025


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

form

solid

mp

70 °C (External MSDS)

storage temp.

15-25°C

SMILES string

O(C(c1ccccc1)C(=O)O)C

InChI

1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)

InChI key

DIWVBIXQCNRCFE-UHFFFAOYSA-N

Analysis Note

Identity (IR-spectrum): conforms
Appearance: White to almost white crystals or crystalline powder.
Melting point: 68 - 72 °C

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service