Skip to Content
Merck
All Photos(1)

Key Documents

1.51238

Supelco

PVDF double cones for HPLC Columns

1/16 in. connector (with knurled nut)

Sign Into View Organizational & Contract Pricing

Select a Size

10 UNITS
CHF 206.00

CHF 206.00


Estimated to ship on19 May 2025



Select a Size

Change View
10 UNITS
CHF 206.00

About This Item

UNSPSC Code:
41115716
NACRES:
SB.52

CHF 206.00


Estimated to ship on19 May 2025


Product Name

PVDF double cones, 1/16 in. connector (with knurled nut)

technique(s)

HPLC: suitable

Quality Level

fitting

1/16 in. connector (with knurled nut)

storage temp.

no temp limit

General description

The package contains: 10 PVDF double cones for capillary tubing (1/16") with knurled screw Ordering No. 1.15545 resp. 1.51216.
for capillary connections 1/16" with knurled nut

Analysis Note

visual control (for delivery): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service