Skip to Content
Merck
All Photos(1)

Key Documents

14-787-M

Sigma-Aldrich

PI3 Kinase (p110α (H1047R)/p85α) Murine, 10 µg

Complex of N-terminal 6His-tagged, recombinant, full-length mouse p110α containing the mutation H1047R α & untagged, recombinant, full length mouse p85α. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 432.00

CHF 432.00


Estimated to ship on12 April 2025



Select a Size

Change View
10 μG
CHF 432.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32

CHF 432.00


Estimated to ship on12 April 2025


biological source

mouse

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 129 kDa (p110α; H1047R)
Mw 83.6 kDa (p85α)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

mouse ... PIK3CA(18706)

General description

Complex of N-terminal 6His-tagged, recombinant, full-length mouse p110α containing the mutation H1047R αand untagged, recombinant, full length mouse p85α.
Product Source: Coexpressed by baculovirus in Sf21 insect cells.
The H1047R substitution in p110α is associated with a number of human cancers. The residue is conserved between the human and mouse sequences. In vitro studies have shown that this mutation confers increased lipid kinase activity compared to wild type, and is able to induce oncogenic transformation.

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: PI3Kα/p85α
Target Sub-Family: PI3K/PI4K

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-787M when ordering the 250μg size.

Physical form

Ni2+/NTA-agarose

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High frequency of mutations of the PIK3CA gene in human cancers.
Yardena Samuels et al.
Science (New York, N.Y.), 304(5670), 554-554 (2004-03-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service