Skip to Content
Merck
All Photos(1)

Key Documents

14-739-M

Sigma-Aldrich

TLK2 Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, 388-end, human TLK2. For use in Kinase Assays.

Synonym(s):

tousled-like kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 411.00
250 μG
CHF 3’310.00

CHF 411.00


Estimated to ship on08 May 2025



Select a Size

Change View
10 μG
CHF 411.00
250 μG
CHF 3’310.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

CHF 411.00


Estimated to ship on08 May 2025


biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 48.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal 6His-tagged, recombinant, 388-end, human TLK2.
Product Source: Expressed by baculovirus in Sf21 cells.

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: TLK2
Target Sub-Family: Misc

Packaging

Also available in 250μg size as catalog # 14-739M - Call for pricing and availability.

Physical form

Ni2+/NTA-agarose

Storage and Stability

Stable for 6 months at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service