Skip to Content
Merck
All Photos(1)

Key Documents

14-723-M

Sigma-Aldrich

CaM Kinase IIδ Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, human, full length CaM Kinase IIδ, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 519.00
250 μG
CHF 3’290.00

CHF 519.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 519.00
250 μG
CHF 3’290.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

CHF 519.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 58 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... CAMK2D(817)

General description

N-terminal 6His-tagged, recombinant, human, full length CaM Kinase IIδ

N-terminal 6His-tagged, recombinant, human, full length CaM Kinase IIδ
Product Source: expressed by baculovirus in Sf21 cells

Application

Research Category
Metabolism

Inflammation & Immunology
Research Sub Category
Obesity

Metabolic Disorders

Osteoporosis

Arthritis

Biochem/physiol Actions

Protein Target: CaMKIIδ
Target Sub-Family: CAMK

Quality

routinely evaluated by phosphorylation of 250μM ZIPtide (Catalog # 12-545)

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service