Skip to Content
Merck
All Photos(1)

Key Documents

14-646-M

Sigma-Aldrich

TCPTP Protein, 10 µg

Un-tagged, recombinant, human TCPTP, amino acids 1-341. For use in Phosphatase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 414.00

CHF 414.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 414.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

CHF 414.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

human

Quality Level

100
300

recombinant

expressed in E. coli

mol wt

Mw 39.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

UniProt accession no.

Gene Information

human ... PTPN2(5771)

General description

Product Source: Human TCPTP, expressed in E. coli cells.
Un-tagged, recombinant, human TCPTP, amino acids 1-341.

Biochem/physiol Actions

Protein Target: TCPTP

Quality

Routinely evaluated by dephosphorylation of DiFMUP.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service