Skip to Content
Merck
All Photos(1)

Key Documents

1.46219

Millipore

Mossel Broth

ready-to-use, tube volume 10 mL , filling volume

Synonym(s):

Buffered Glucose-Brilliant Green Bile Broth, EE Broth, Enterobacteriaceae Enrichment Broth acc. to Mossel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP
USP

Quality Level

description

Selective enrichment of Enterobacteriaceae from non-sterile pharmaceutical products, foodstuff, and other materials

form

liquid (ready-to-use)

feature

closure type screw cap with septum
ready-to-use

packaging

single packed of (10 ml in 17 ml tube with yellow cap (20 or 100 tubes per box))
tube of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

tube capacity

17 mL

tube volume

10 mL , filling volume

pH

7.2

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

enterobacteriaceae

General description

Mossel Broth is a selective enrichment media for bile-tolerant, Gram-negative bacteria, and particularly Enterobacteriaceae such as Salmonella, Shigella, and other enteric pathogens from non-sterile pharmaceutical products, foodstuff, and other materials. Glucose is the main energy source and supports the rapid growth of all Enterobacteriaceae. The enzymatic digest of animal tissue provides nitrogen, vitamins, and amino acids. The high buffering capacity of the culture medium prevents the acid formed from killing the culture. The undesired accompanying bacterial flora is completely inhibited by brilliant green and bile.

Application

Mossel Broth is designed for selective enrichment of bile-tolerant, Gram-negative bacteria, particularly Enterobacteriaceae such as Salmonella, Shigella and other enteric pathogens from non-sterile pharmaceutical products, foodstuff, and other materials.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service