Skip to Content
Merck
All Photos(1)

Key Documents

14-607-M

Sigma-Aldrich

PIM2 Protein, active, 25 µg

Active, human PIM2 expressed in E. coli, purified using GSH agarose, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 519.00

CHF 519.00


Estimated to ship on20 May 2025



Select a Size

Change View
25 μG
CHF 519.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

CHF 519.00


Estimated to ship on20 May 2025


biological source

human

Quality Level

mol wt

Mw 63.4 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PIM2(11040)

General description

Product Source: N-terminal GST-tagged recombinant full-length human PIM2
human PIM2 expressed in E. coli, purified using GSH agarose

Quality

routinely evaluated by phosphorylation of PIM2tide

Physical form

GSH agarose

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casey J Fox et al.
Genes & development, 17(15), 1841-1854 (2003-07-19)
Growth factor withdrawal results in the termination of factor-dependent transcription. One transcript that declines rapidly following growth factor deprivation of hematopoietic cells is the serine/threonine kinase pim-2. When constitutively expressed, Pim-2 conferred long-term resistance to a variety of apoptotic stimuli
Bin Yan et al.
The Journal of biological chemistry, 278(46), 45358-45367 (2003-09-05)
Hematopoietic growth factors mediate the survival and proliferation of blood-forming cells, but the mechanisms through which these proteins produce their effects are incompletely known. Recent studies have identified the pim family of kinases as mediators of cytokine-dependent survival signals. Several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service