Skip to Content
Merck
All Photos(1)

Key Documents

1.44256

Millipore

Docking Station

for use with RCS® High Flow Touch, for convenient charging

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 707.00

CHF 707.00


Estimated to ship on26 May 2025


Request more information

Select a Size

Change View
1 EA
CHF 707.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

CHF 707.00


Estimated to ship on26 May 2025


Request more information

material

polycarbonate stand

Agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

room temp

General description

Docking Station is an additional accessory for use with RCS® High Flow Touch air sampler. The RCS® High Flow Touch is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. The Docking station used for recharging the integrated Lithium-ion battery of the RCS® High Flow Touch Microbial Air Sampler.

Application

Docking Station is an additional accessory with the RCS® microbial air sampler used for recharging the integrated Lithium-ion battery. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service