Skip to Content
Merck
All Photos(1)

Key Documents

1.44150

Millipore

Carrying Case

for use with RCS® High Flow Touch

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 405.00

CHF 405.00


Estimated to ship on14 May 2025



Select a Size

Change View
1 EA
CHF 405.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

CHF 405.00


Estimated to ship on14 May 2025


material

aluminum case

Agency

non-compendial

description

stability

feature

portable

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

shipped in

ambient

storage temp.

room temp

General description

Carrying Case is an additional accessory for use with RCS® High Flow Touch microbial air sampler. The RCS® High Flow Touch is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. This microbial air sampler is based on impaction by centrifugal force and can also be used with a compressed gas adapter for additional monitoring of compressed gases The carrying case is made of aluminum and have sections for the RCS® High Flow Touch Microbial Air Sampler, two rotors, power supply, RS232 serial cable, USB adapter, documents and agar strips.

Application

Carrying Case is an additional accessory with the RCS® High Flow air sampler used safety transportation of the air sampler. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service