Skip to Content
Merck
All Photos(1)

Key Documents

14-252-M

Sigma-Aldrich

p38α/SAPK2a Protein, unactive, 50 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

mol wt

Mw 67.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

General description

Recombinant GST fusion protein containing full-length human p38α/SAPK2a
expressed in E. Coli

Application

Unactive, recombinant GST fusion protein containing full-length human p38α/SAPK2a, for use in Kinase Assays.

Biochem/physiol Actions

  • Protein Target: p38α/SAPK2a
  • Target Sub-Family:CMGC

Quality

routinely evaluated by phosphorylation of MBP

Storage and Stability

6 months at -20 C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service