132233
Fluoride Standard Solution
traceable to SRM from NIST 0.50 mg/l F in H₂O
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
liquid
specific analyte(s)
fluoride
technique(s)
photometry: suitable
pH
4.7 ( in H2O)
density
1.00 g/cm3 at 20 °C
shipped in
ambient
storage temp.
15-25°C
General description
This Spectroquant® Fluoride Standard Solution is a high-quality laboratory solution used for internal quality control and method-controlling purposes.
These Spectroquant® standard solutions perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.
Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.
These Spectroquant® standard solutions perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.
Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.
Application
Fluoride Standard Solution can be used to check the photometric measurement system (reagents, measurement device, handling) and the mode of working.
Features and Benefits
- Manufactured and tested in DIN EN ISO 9001:2008 accredited laboratory
- The solution is pre-diluted to a concentration that falls into the range of our Spectroquant® test kits. This eliminates the need for dilution prior to analysis and hence avoids a common source of error
- Ready-to-use diluted reference materials (RMs) along with expanded measurement uncertainty ensure absolute precision
- Accompanied with a certificate of analysis (COA) detailing the certified values, including uncertainty and expiry date
- Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST) or to United States Pharmacopeia (USP)
- Reliable and reproducible results
Analysis Note
Concentration: 0.50 ± 0.02 mg/L F in H2O
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service