1.25159
Eshmuno® CPX Resin
RoboColumn Eshmuno® CPX, volume 0.6 mL, prepacked column
Synonym(s):
RoboColumn Eshmuno® CPX 0.6ml
About This Item
ligand
(Sulfoisobutyl)
Quality Level
material
polypropylene housing
product line
Eshmuno®
form
prepacked column
manufacturer/tradename
Eshmuno®
parameter
≤8 bar pressure
500 cm/hr max. flow rate
column diam. × L
5 mm × 30 mm
volume
0.6 mL
matrix active group
Polyvinyl Ether
mean particle size
50 μm
capacity
~120 mL, resin binding capacity
application(s)
mAb
recombinant protein
vaccine development
separation technique
cation exchange
storage temp.
no temp limit
General description
Features and Benefits
- Intermediate purification or polishing of monoclonal antibodies
- High aggregate removal efficiency combined with high dynamic binding capacity
- High loading capacity combined with high purity at lower elution volumes.
- Wide pH and conductivity operating window
- High flow rate versus pressure-flow behavior
- Excellent removal of impurities
- Robust and safe packing procedures
- Strong chemical stability
- Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH
Legal Information
related product
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service