Skip to Content
Merck

1.20093

Millipore

Eshmuno® CP-FT Resin

Synonym(s):

Eshmuno® CP-FT

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
CHF 155.00
100 ML
CHF 1’319.00
500 ML
CHF 4’666.00
5 L
Inquire

CHF 155.00


Estimated to ship on01 June 2025



Select a Size

Change View
10 ML
CHF 155.00
100 ML
CHF 1’319.00
500 ML
CHF 4’666.00
5 L
Inquire

About This Item

UNSPSC Code:
41115711

CHF 155.00


Estimated to ship on01 June 2025


Quality Level

ligand

(Sulfoisobutyl)

sterility

sterile (Caustic Stable)

product line

Eshmuno®

form

resin

specific analyte(s)

MAbs

parameter

400 cm/hr flow rate
8 bar max. pressure

matrix active group

polymer, polyvinyl ether

mean particle size

50 μm

capacity

70 mg binding capacity (lysozyme/ml of resin)

transition temp

flash point 35 °C

separation technique

cation exchange
strong cation exchange

storage temp.

2-30°C

General description

Cation exchange resin specifically developed for the flow-through removal of aggregates using frontal chromatography

Features and Benefits

Eshmuno® CP-FT chromatography resin enables:

  • Superior flow-through removal of mAb aggregates
  • High product recoveries at high mass loadings
  • Significant reduction in resin and buffer volume
  • Smaller manufacturing footprint (smaller columns, buffer tanks, etc.)
  • Low salt process conditions eliminate the need for dilution before subsequent ion exchange steps
  • Significant reduction in processing volumes improves virus filtration and ultrafiltration processing economics
  • Rigid base bead enables higher flow rates and easier column packing
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

  • 1.20093.0100: Eshmuno® CP-FT Resin 100 ml
  • 1.20093.0010: Eshmuno® CP-FT Resin 10 ml
  • 1.20093.0500: Eshmuno® CP-FT Resin 500 ml
  • 1.20093.5000: Eshmuno® CP-FT Resin 5 L
Plastic bottle

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service