Skip to Content
Merck
All Photos(1)

Key Documents

1.19247

Supelco

Ruthenium(III) chloride hydrate

for analysis EMSURE®

Synonym(s):

Ruthenium(III) chloride hydrate, Ruthenium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl3Ru · xH2O
CAS Number:
Molecular Weight:
207.43 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-167-5
form:
powder

Quality Level

product line

EMSURE®

form

powder

concentration

35.0-40.0% Ru

pH

<2 (20 °C in H2O, Aqueous solution)

density

3.11 g/cm3

cation traces

Ag: ≤50 ppm
Al: ≤10 ppm
Au: ≤100 ppm
Ca: ≤50 ppm
Cd: ≤10 ppm
Co: ≤10 ppm
Cr: ≤50 ppm
Cu: ≤50 ppm
Fe: ≤100 ppm
Hg: ≤100 ppm
Ir: ≤100 ppm
K: ≤50 ppm
Mg: ≤50 ppm
Mn: ≤10 ppm
Na: ≤100 ppm
Ni: ≤10 ppm
Os: ≤100 ppm
Pb: ≤50 ppm
Pd: ≤100 ppm
Pt: ≤100 ppm
Rh: ≤100 ppm
Zn: ≤10 ppm

storage temp.

15-25°C

Analysis Note

Identity (elemental analysis): passes test
Assay (Ru): 35.0 - 40.0 %
Ag (Silver): ≤ 50 ppm
Al (Aluminium): ≤ 10 ppm
Au (Gold): ≤ 100 ppm
Ca (Calcium): ≤ 50 ppm
Cd (Cadmium): ≤ 10 ppm
Co (Cobalt): ≤ 10 ppm
Cr (Chromium): ≤ 50 ppm
Cu (Copper): ≤ 50 ppm
Fe (Iron): ≤ 100 ppm
Hg (Mercury): ≤ 100 ppm
Ir (Iridium): ≤ 100 ppm
K (Potassium): ≤ 50 ppm
Mg (Magnesium): ≤ 50 ppm
Mn (Manganese): ≤ 10 ppm
Na (Sodium): ≤ 100 ppm
Ni (Nickel): ≤ 10 ppm
Os (Osmium): ≤ 100 ppm
Pb (Lead): ≤ 50 ppm
Pd (Palladium): ≤ 100 ppm
Pt (Platinum): ≤ 100 ppm
Rh (Rhodium): ≤ 100 ppm
Zn (Zinc): ≤ 10 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service