Skip to Content
Merck
All Photos(1)

Key Documents

119123

Millipore

Adenosine 5′-O-(3-Thiotriphosphate), Solution, Tetralithium Salt

A phosphatase-resistant ATP analogue.

Synonym(s):

Adenosine 5′-O-(3-Thiotriphosphate), Solution, Tetralithium Salt, ATP-γ-S, 4Li

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 341.00

CHF 341.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 341.00

About This Item

Empirical Formula (Hill Notation):
C10H12N5O12P3S · 4Li
CAS Number:
Molecular Weight:
546.98
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

CHF 341.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

>90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

<10% ADP (HPLC)

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].[P](=O)([S-])([O-])O[P](=O)([O-])O[P](=O)([O-])OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n]2c3ncnc(c3nc2)N

InChI

1S/C10H16N5O12P3S.4Li/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(25-10)1-24-28(18,19)26-29(20,21)27-30(22,23)31;;;;/h2-4,6-7,10,16-17H,1H2,(H,18,19)(H,20,21)(H2,11,12,13)(H2,22,23,31);;;;/q;4*+1/p-4/t4-,6-,7-,10-;;;;/m1..../s1

InChI key

DWQFDOIBOYDYKH-KWIZKVQNSA-J

General description

A phosphatase-resistant ATP analog that is reported to rearrange F-actins into membrane ruffles.
A phosphatase-resistant ATP analogue.

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a 100 mM aqueous solution in water, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Ludowyke, R.I., et al. 1994. Eur. J. Cell Biol.64, 357.
Shimizu, T., et al. 1989. Biochemistry28, 7022.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service