Skip to Content
Merck
All Photos(1)

Key Documents

1.18466

Supelco

Phosphate Test Refill pack

for use with MQUANT®

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
CHF 170.00

CHF 170.00


Estimated to ship on18 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
CHF 170.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

CHF 170.00


Estimated to ship on18 May 2025


Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

phosphate

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kits 1.18388 and 1.14449 for the colorimetric evaluation of phosphate. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical and ecological system.

Method
In sulfuric solution orthophosphate ions react with ammonium vanadate and ammonium heptamolybdate to form orange-yellow molybdovanadophosphoric acid. The phosphate concentration is measured semiquantitatively by visual colorimetric evaluation.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service