Skip to Content
Merck
All Photos(1)

Documents

1.18457

Supelco

Cyanide Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Cyanide Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

feature

refill

Quality Level

specific analyte(s)

cyanide

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all reagents for the MQuant® test kits 1.14798, 1.14429, and 1.14417 for the colometric evaluation of cyanide.
for 114798, 114429, 114417 MQuant®

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service