Skip to Content
Merck
All Photos(1)

Key Documents

1.18338

Supelco

Lamp

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
CHF 447.00

CHF 447.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
CHF 447.00

About This Item

UNSPSC Code:
47101516
NACRES:
SE.11

CHF 447.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

technique(s)

photometry: suitable

compatibility

for use with Turbiquant 1500 T
for use with Turbiquant 3000 T

storage temp.

2-30°C

General description

1500 T, 3000 T Turbiquant
The lamp in the Turbiquant 1500 T and 3000 T can be replaced if necessary. Only use the lamp that is intended for use with your instrument. For other turbidimeters please choose the respective lamp.

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service