Key Documents
1.17340
Millipore
MAS-100 Atmos® Dust Cover
for use with MAS-100 Atmos®
Select a Size
1173400001
CHF 86.70
CHF 86.70
Estimated to ship on01 June 2025
Request more information
Select a Size
Change View
1173400001
CHF 86.70
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
CHF 86.70
Estimated to ship on01 June 2025
Request more information
material
white polypropylene cover
description
Compressed gas sampling
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
MAS-100 Atmos® Dust Cover is an accessory to protect the instrument from dust. If the perforated lid is dismantled for autoclaving, it is recommended to protect the device with the MAS-100 Atmos® dust cover.
The dust cover is white and made from polypropylene. It can be sterilized by steam sterilization (autoclaving) at 134°C for 5 minutes or 121°C for 20 minutes. Disinfection is done by spraying or wiping with 70% Ethanol or 70% IPA or other disinfectants compatible with polypropylene.
Application
The MAS-100 Atmos® Dust Cover is used to prevent dust accumulation on the sampling head of MAS-100 Atmos® Microbial Compressed Gas Sampler.
Analysis Note
The MAS-100 Atmos® dust cover will slightly shrink after the first autoclaving cycle, resulting in some clearance. This should be considered during transport but has no effect on the protective capacity of the cover.
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG