Key Documents
1.17278
Millipore
HEPA H13 Filter
74 mm, Exchange filter, for use with MAS-100 NT® (with filter), for use with MAS-100 Atmos®
Select a Size
1 EACH
CHF 75.90
CHF 75.90
Estimated to ship on02 May 2025
Request more information
Select a Size
Change View
1 EACH
CHF 75.90
About This Item
UNSPSC Code:
41103403
CHF 75.90
Estimated to ship on02 May 2025
Request more information
material
polypropylene filter
Agency
non-compendial
feature
filter
manufacturer/tradename
MAS-100 NT®
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
for use with MAS-100 NT® (with filter)
General description
The HEPA H13 Filter 74 mm for MAS-100 NT/NT Ex is an replacement filter for MAS-100 NT® w. filter, MAS-100 NT® Ex w. filter as well as for MAS-100 NT® or MAS-100 NT® Ex which are upgraded with the mounting Kit for Filter. The filter should be replaced on yearly bases. To ease the workflow, it is rcommended to replace the filter along with the yearly calibration/adjustment of the microbial air sampler.
Application
The HEPA H13 Filter is a replacement filter for microbial air samplers MAS-100 NT® with filter and MAS-100 NT® Ex with filter. Microbial air sampling devices are extensively used in Pharmaceutical applications.
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG