Skip to Content
Merck
All Photos(1)

Key Documents

1.17244

Supelco

Cobalt Test, photometric

photometric, 0.05-2.00 mg/L (Co2+), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
CHF 147.00

CHF 147.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
CHF 147.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

CHF 147.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Product Name

Cobalt Cell Test, photometric, 0.05-2.00 mg/L (Co2+), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

cobalt

measuring range

0.05-2.00 mg/L (Co2+)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Nitroso-R salt)

storage temp.

15-25°C

General description

Method: photometric 0.05 - 2.00 mg/l Co Spectroquant®
Cobalt(II) ions react with Nitroso-R salt to form a orange-red complex that is determined photometrically. Our Spectroquant® Cobalt Test allows you to accurately quantify the Co levels in your sample.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) , which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service