Skip to Content
Merck
All Photos(1)

Documents

1.14410

Supelco

Silicate (Silicic Acid) Test Kit, colorimetric

0.01-0.25 mg/L (Si), 0.02-0.53 mg/L (SiO₂), MQuant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Silicate (Silicic Acid) Test, colorimetric, 0.01-0.25 mg/L (Si), 0.02-0.53 mg/L (SiO2), for use with MCOLORTEST®

specific analyte(s)

silicate

Quality Level

measuring range

0.01-0.25 mg/L (Si)
0.02-0.53 mg/L (SiO2)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.01-0.02-0.04-0.06-0.08-0.10-0.15-0.20-0.25 mg/l Si 0.02-0.04-0.09-0.13-0.17-0.21-0.32-0.43-0.53 mg/l SiO₂ MColortest®
The MQuant® Silicate (Silicic Acid) test (formally MColortest®) can be used for the determination of silicate in e.g.:
  • Groundwater, surface water, and seawater
  • Drinking water and mineral water
  • Industrial and process water
  • Boiler water and boiler feed water
  • Wastewater and percolating water

Method
In sulfuric solution silicate ions react with molybdate ions to form a yellow heteropoly acid. This is reduced to silicomolybdenum blue. The silicate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18323 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Analysis Note

Application testconforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service