Skip to Content
Merck
All Photos(1)

Documents

1.12220

Supelco

Lanthanum(III) oxide

EMPLURA®

Synonym(s):

Lanthanum(III) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La2O3
CAS Number:
Molecular Weight:
325.81
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-200-5
NACRES:
NA.21

Quality Level

product line

EMPLURA®

Assay

≥99.5% (the ignited basis substance, complexometric)

form

solid

potency

>9968 mg/kg LD50, oral (Rat)

pH

9.0 (20 °C, 50 g/L in H2O, slurry)

mp

2315 °C

density

6.51 g/cm3 at 20 °C

bulk density

1300 kg/m3

storage temp.

no temp limit

InChI

1S/2La.3O/q2*+3;3*-2

InChI key

MRELNEQAGSRDBK-UHFFFAOYSA-N

Related Categories

Linkage

Replaces: LX0045-2; LX0045

Analysis Note

Assay (complexometric, on of the ignited basis substance): ≥ 99.5 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service