Skip to Content
Merck
All Photos(1)

Key Documents

1.10294

Millipore

Enterococci broth

for microbiology Chromocult®

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CHF 644.00

CHF 644.00


Estimated to ship on21 May 2025


Request more information

Select a Size

Change View
500 G
CHF 644.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

CHF 644.00


Estimated to ship on21 May 2025


Request more information

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.3-7.7 (25 °C, 17.8 g/L in H2O, after autoclaving)

solubility

17.8 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Enterococcus spp.

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish
pH-value (25 °C): 7.3 - 7.7
Growth (Enterococcus faecalis ATCC 11700): fair to very good
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): fair to very good
Growth (Enterococcus faecium ATCC 6057): fair to very good
Growth (Enterococcus durans ATCC 19432): fair to very good
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): none to very good
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): none to poor
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): none to poor
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): none to poor
colour change to blue-green (Enterococcus faecalis ATCC 11700): +
colour change to blue-green (Enterococcus faecalis ATCC 19433 (WDCM 00009)): +
colour change to blue-green (Enterococcus faecium ATCC 6057): +
colour change to blue-green (Enterococcus durans ATCC 19432): +
colour change to blue-green (Staphylococcus aureus ATCC 25923 (WDCM 00034)): -
colour change to blue-green (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
colour change to blue-green (Escherichia coli ATCC 25922 (WDCM 00013)): -
colour change to blue-green (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Incubation: 24 - 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service