1.09450
pH-Indicator strips pH 2.0 - 9.0
non-bleeding, colorimetric, individually sealed, MQuant®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
pH-Indicator strips pH 2.0 - 9.0, non-bleeding, colorimetric, pH range 2.0-9.0, MQuant®
product line
MQuant®
Quality Level
feature
non-bleeding
pH range
2.0-9.0
detection method
colorimetric
storage temp.
2-30°C
Related Categories
General description
The pH tests are suited for all media in environmental analysis and in industrial in-process controls. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it.
individually sealed MColorpHast®
Legal Information
MCOLORPHAST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service