Skip to Content
Merck
All Photos(1)

Key Documents

1.09196

Supelco

Alkali blue (C.I. 42765)

passes application test for DIN 51558

Synonym(s):

Alkali blue (C.I. 42765)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 70.50

CHF 70.50


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
CHF 70.50

About This Item

Empirical Formula (Hill Notation):
C37H29N3O3S
CAS Number:
Molecular Weight:
595.71
MDL number:
UNSPSC Code:
12171503
EC Index Number:
215-385-2
NACRES:
NA.21

CHF 70.50


Estimated to ship on29 May 2025


Request a Bulk Order

Agency

suitable for DIN 51558

Quality Level

form

solid

loss

≤5% loss on drying, 110°C

visual transition interval

11.5-13, blue-violet to pink

λmax

597-602 nm in ethanol

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])c1c(cccc1)[N+H]=C2C=CC(=C(c5ccc(cc5)Nc6ccccc6)c3ccc(cc3)Nc4ccccc4)C=C2

InChI

1S/C37H29N3O3S/c41-44(42,43)36-14-8-7-13-35(36)40-34-25-19-29(20-26-34)37(27-15-21-32(22-16-27)38-30-9-3-1-4-10-30)28-17-23-33(24-18-28)39-31-11-5-2-6-12-31/h1-26,38-39H,(H,41,42,43)

InChI key

DVPLSZDCYOJSOG-UHFFFAOYSA-N

General description

indicator

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Transition range: pH 11.5 - pH 13.0 blue-violet - dark-pink
Absorption maximum λmax. (Ethanol): 597 - 602 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol abs.; calculated on dried substance): ≥ 1000
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability as indicator (according to DIN 51558): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service