Skip to Content
Merck
All Photos(2)

Documents

1.08556

Supelco

Hydrogen peroxide solution

35%, EMPLURA®

Synonym(s):

Hydrogen peroxide solution, Perhydrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
NACRES:
NA.21

product name

Hydrogen peroxide 35%, EMPLURA®

vapor pressure

20 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

Quality Level

product line

EMPLURA®

form

liquid

potency

1193-1270 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

concentration

≥35.0% (manganometric)

impurities

≤0.005% Chloride (Cl)

pH

2-4 (20 °C in H2O)

bp

110 °C/1013 hPa

mp

-24 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • An In Vitro Study on the Effects of Hydrogen Peroxide-Based Bleaching Agents on Enamel: This study used Field Emission Scanning Electron Microscopy (FESEM) with Energy Dispersive Spectroscopy (EDS) to evaluate the impact of hydrogen peroxide-based bleaching agents on enamel, emphasizing the structural changes induced by 35% hydrogen peroxide (Ps S et al., 2024).

Analysis Note

Assay (manganometric): ≥ 35.0 %
Free acid (as sulfuric acid): ≤ 0.2
Chloride (Cl): ≤ 0.005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service