Skip to Content
Merck
All Photos(1)

Key Documents

1079

Millipore

N-Acetyl-D-glucosamine

An acceptor substrate for galactosyltransferases. Inhibits the lectin WGA and β-N-acetylhexosaminidase (Ki = 430 µM). When added to a medium, it improves the recovery of Haemophilus influenza from sputum.

Synonym(s):

N-Acetyl-D-glucosamine, NAG, GlcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white to beige
white to off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(C1C(OC(C(C1O)O)CO)O)C(=O)C

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)

InChI key

OVRNDRQMDRJTHS-UHFFFAOYSA-N

General description

Acceptor substrate for galactosyltransferases. Lectin inhibitory amino sugar. When added to the medium, it improves the recovery of Haemophilus influenza from sputum. A competitive inhibitor of β-N-acetylhexosaminidase (Ki = 430 µM).
An acceptor substrate for galactosyltransferases. Inhibits the lectin WGA and β-N-acetylhexosaminidase (Ki = 430 µM). When added to a medium, it improves the recovery of Haemophilus influenza from sputum.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Sakai, K., et al. 1994. Appl. Environ. Microbiol. 60, 2911.
Moller, L.V., et al. 1993. J. Clin. Microbiol. 31, 1952.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Sakai et al.
Applied and environmental microbiology, 60(8), 2911-2915 (1994-08-01)
Thermostable exochitinase was purified to homogeneity from the culture fluid of Bacillus stearothermophilus CH-4, which was isolated from agricultural compost containing shrimp and crabs. The enzyme was a single polypeptide with a molecular mass of 74 kDa, and the N-terminal
L V Möller et al.
Journal of clinical microbiology, 31(7), 1952-1954 (1993-07-01)
A modified selective medium supplemented with N-acetyl-D-glucosamine (NAG), hemin, and NAD plus two cefsulodin disks, for primary isolation of nonencapsulated Haemophilus influenzae from sputum of patients with cystic fibrosis, is described. Isolation of H. influenzae from this medium, designated NAG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service