Skip to Content
Merck
All Photos(2)

Key Documents

1.06566

Supelco

Sodium phosphate dibasic

anhydrous 99.99 Suprapur®

Synonym(s):

Sodium phosphate dibasic, di-Sodium hydrogen phosphate, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CHF 331.00
500 G
CHF 1’460.00

CHF 331.00


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
100 G
CHF 331.00
500 G
CHF 1’460.00

About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7
NACRES:
NB.21
Assay:
99.99%
grade:
for inorganic trace analysis
form:
solid
solubility:
77 g/L

CHF 331.00


Estimated to ship on26 March 2025


Request a Bulk Order

grade

for inorganic trace analysis

Quality Level

Assay

99.99%

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

8.7-9.3 (20 °C, 10 g/L in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

250 °C (decomposition)

solubility

77 g/L

bulk density

880 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (acidimetric): ≥ 99.0 %
Absorbance (at 260 nm; 10 %; 2 cm; water): ≤ 0.010
Chloride (Cl): ≤ 5 ppm
Sulfate (SO₄): ≤ 30 ppm
Total nitrogen (N): ≤ 10 ppm
As (Arsenic): ≤ 0.1 ppm
Ba (Barium): ≤ 10.0 ppm
Ca (Calcium): ≤ 0.50 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.050 ppm
Cs (Cesium): ≤ 5 ppm
Cu (Copper): ≤ 0.010 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
Hg (Mercury): ≤ 0.05 ppm
K (Potassium): ≤ 2.0 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 1.0 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.010 ppm
Ni (Nickel): ≤ 0.050 ppm
Pb (Lead): ≤ 0.010 ppm
Rb (Rubidium): ≤ 5.0 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.50 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.050 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service