Skip to Content
Merck

1.06444

SAFC

Sodium chloride

EMPROVE® API Ph Eur,BP,JP,USP

Pharma Manufacturing

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-598-3
Pricing and availability is not currently available.

Agency

BP
JP
Ph. Eur.
USP

Quality Level

vapor pressure

1.3 hPa ( 865 °C)

product line

EMPROVE® API

form

solid

potency

3000 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C

mp

801 °C (lit.)

solubility

358 g/L

bulk density

1140 kg/m3

application(s)

liquid formulation
ophthalmics
parenterals
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-25°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium chloride is used in ophthalmic, dialysis, clinical nutrition and infusion solutions. Sodium chloride Emprove® API is manufactured according to ICH Q7 guidelines. A CEP is available.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Elemental Impurities Information

Available by subscription or for a fee

API Information Package

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service