Skip to Content
Merck
All Photos(1)

Key Documents

1.05570

Supelco

TLC Silica gel 60 F₂₅₄

pkg of 25 sheets, sheet L × W 20 cm × 10 cm, aluminum support

Sign Into View Organizational & Contract Pricing

Select a Size

25 UNITS
CHF 157.00

CHF 157.00


Estimated to ship on26 May 2025



Select a Size

Change View
25 UNITS
CHF 157.00

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

CHF 157.00


Estimated to ship on26 May 2025


material

aluminum support
silica gel 60 matrix

Quality Level

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 25 sheets

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

200 μm

plate L × W

5 cm × 20 cm

sheet L × W

20 cm × 10 cm

particle size

10-12 μm

pore size

60 Å medium pore diameter

storage temp.

2-30°C

General description

25 Aluminium sheets 10 x 20 cm
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.

Application


  • Nonstoichiometric Protic Ionic Liquids: The Role of Excess Acid in Charge Transport Mechanisms: This research examines the influence of trifluoromethanesulfonic acid in creating nonstoichiometric protic ionic liquids, providing a foundation for enhancing ionic conductivity and battery technologies (Middendorf and Schönhoff, 2024).

  • B(9)-OH-o-Carboranes: Synthesis, Mechanism, and Property Exploration: The study explores the utilization of trifluoromethanesulfonic acid in the synthesis of boron-rich clusters, highlighting its role in the development of new materials with unique chemical properties (Ma et al., 2023).

  • High pressure as a novel tool for the cationic ROP of γ-butyrolactone: Demonstrates trifluoromethanesulfonic acid′s application in polymer chemistry, specifically in ring-opening polymerization under high-pressure conditions, opening avenues for materials science research (Bernat et al., 2021).

  • Peptide Cyclization Mediated by Metal-Free S-Arylation: This research utilizes trifluoromethanesulfonic acid to facilitate peptide cyclization, a crucial reaction in the development of therapeutic peptides and pharmaceutical applications (Kobayashi et al., 2021).



Analysis Note

Specific surface area (according to BET; 5-Pt. measurement): 480 - 540 m²/g
Pore volume (N₂-isotherm): 0.74 - 0.84 ml/g
d 50 (laser diffraction, size distribution): 9.5 - 11.5 µm
Layer thickness: 165 - 235 µm
Deviation of layer thickness per plate: ≤ 35 µm
Chromatographic testing:
colour test
hRf-values
- bleu vif organol, colour test, lipophile:
11 - 25
- Ceres Black G, colour test, lipophile: 34 - 48
- ceres Violet brn, colour test, lipophile: 52 - 67
Typical value determined on a conditioned sheet
Eluent: toluene (45% rel. humidity)

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service