Skip to Content
Merck
All Photos(1)

Key Documents

1.05217

Supelco

Copper di-ammonium Titriplex® solution

Cu(NH₄)₂-EDTA 0.1 mol/l

Synonym(s):

Cooper di-ammonium EDTA solution; Cooper di-ammonium ethylenediaminetetraacetic acid solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
CHF 195.00

CHF 195.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
500 ML
CHF 195.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.21

CHF 195.00


Estimated to ship on29 May 2025


Request more information

Quality Level

form

liquid

reaction suitability

reaction type: Acid-base reactions
reaction type: Complexometric reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

9.7-10.3 ( in H2O)

density

1.02 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Assay (Cu): 0.098 - 0.102 mol/l
pH-value: 9.7 - 10.3
Free EDTA: conforms
Free copper: conforms

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service