Skip to Content
Merck
All Photos(1)

Key Documents

1.03023

Supelco

Bromophenol red

pH indicator

Synonym(s):

Bromophenol red, 3′,3′′-Dibromophenolsulfonphthalein, BPR

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 354.00

CHF 354.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
CHF 354.00

About This Item

Empirical Formula (Hill Notation):
C19H12Br2O5S
CAS Number:
Molecular Weight:
512.17
MDL number:
UNSPSC Code:
12171502
EC Index Number:
220-538-1
NACRES:
NA.21

CHF 354.00


Estimated to ship on21 May 2025


Request a Bulk Order

Product Name

Bromophenol red, indicator

Quality Level

form

solid

impurities

≤5% Water (Karl Fischer)

visual transition interval

4.7-6.3, yellow to purple

mp

230 °C (decomposition)

bulk density

770 kg/m3

Amax

435-440 nm (buffer pH 4.7)
574-577 nm (buffer pH 6.3)

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OC(c4c1cccc4)(c3cc(c(cc3)O)Br)c2cc(c(cc2)O)Br

InChI

1S/C19H12Br2O5S/c20-14-9-11(5-7-16(14)22)19(12-6-8-17(23)15(21)10-12)13-3-1-2-4-18(13)27(24,25)26-19/h1-10,22-23H

InChI key

OYCLSQDXZMROJK-UHFFFAOYSA-N

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Transition range: pH 4.7 - pH 6.3 yellow to purple
Absorption maximum λ 1 (buffer pH 4.7): 435 - 440 nm
Absorption maximum λ 2 (buffer pH 6.3): 574 - 577 nm
Spec. Absorptivity A 1%/1cm (λ1max; 0.01 g/l; buffer pH 4.7; calculated on anhydrous substance): ≥ 380
Spec. Absorptivity A 1%/1cm (λ2max; 0.01 g/l; Puffer pH 6.3; calculated on anhydrous substance): ≥ 760
Water (according to Karl Fischer): ≤ 5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service