Skip to Content
Merck
All Photos(1)

Key Documents

1.02968

Supelco

Neocuproine

GR for analysis

Synonym(s):

Neocuproine, 2,9-Dimethyl-1,10-phenanthroline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 210.00

CHF 210.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
5 G
CHF 210.00

About This Item

Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26
MDL number:
UNSPSC Code:
12352005
EC Index Number:
207-601-9
NACRES:
NA.21

CHF 210.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest more information

Quality Level

form

solid

specific analyte(s)

cyanide

technique(s)

photometry: suitable

mp

159-164 °C

bulk density

280 kg/m3

storage temp.

15-25°C

SMILES string

n1c2c3nc(ccc3ccc2ccc1C)C

InChI

1S/C14H12N2/c1-9-3-5-11-7-8-12-6-4-10(2)16-14(12)13(11)15-9/h3-8H,1-2H3

InChI key

IYRGXJIJGHOCFS-UHFFFAOYSA-N

Analysis Note

Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 99.0 %
Identity (IR-spectrum): conforms
Melting point: 159 - 164 °C
Water (according to Karl Fischer): ≤ 5.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service