Skip to Content
Merck
All Photos(2)

Key Documents

1.02644

SAFC

Collodion solution

4%

Synonym(s):

Cellulose nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
CHF 173.00

CHF 173.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
CHF 173.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352401

CHF 173.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest a Sample

vapor pressure

576 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

170 °C

expl. lim.

1.7-36 % (v/v)

concentration

4%

transition temp

flash point -40 °C

density

0.76 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

15-25°C

SMILES string

[N+](=O)([O-])OC3OC(C(C(C3O[N+](=O)[O-])O[N+](=O)[O-])OC4OC(C(C(C4O[N+](=O)[O-])O[N+](=O)[O-])O[N+](=O)[O-])CO[N+](=O)[O-])CO[N+](=O)[O-].O1C(C(C(C(C1CO)O)O)O)OC2C(OC(C(C2O)O)O)CO

InChI

1S/C12H14N8O27.C12H22O11/c21-13(22)37-1-3-5(7(43-16(27)28)10(46-19(33)34)12(40-3)47-20(35)36)41-11-9(45-18(31)32)8(44-17(29)30)6(42-15(25)26)4(39-11)2-38-14(23)24;13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-12H,1-2H2;3-20H,1-2H2

InChI key

OWWGYSXLINVAES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Collodion 4% is suitable for use in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service