Skip to Content
Merck
All Photos(1)

Key Documents

1.02596

Supelco

Ammonium nitrate solution

1 mol/l acc. to DIN 19730

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
CHF 574.00

CHF 574.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
CHF 574.00

About This Item

UNSPSC Code:
41116107
NACRES:
NB.21

CHF 574.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

liquid

bp

100 °C/1013 hPa

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • Raman Spectroscopy of Glass Beads in Ammonium Nitrate Solution and Compensation of Signal Losses.: This study explores the use of ammonium nitrate solution in Raman spectroscopy to enhance signal detection, demonstrating advancements in analytical techniques for material characterization (Spoor E et al., 2024).

  • Simultaneous speciation analysis of arsenic and iodine in human urine by high performance liquid chromatography-inductively coupled plasma mass spectrometry.: The research utilizes ammonium nitrate solution in the sample preparation process for speciation analysis of arsenic and iodine, showcasing its importance in precise biochemical analysis (Zhou S et al., 2024).

  • Magnesium in dairy cattle nutrition: A meta-analysis on magnesium absorption in dairy cattle and assessment of simple solubility tests to predict magnesium availability from supplemental sources.: This study employs ammonium nitrate solution in solubility tests to predict magnesium availability, contributing to improvements in agricultural practices and animal nutrition (Khiaosa-Ard R et al., 2023).

  • Electromass Transfer in the System "Cation Exchange Membrane-Ammonium Nitrate Solution".: This research investigates the electromass transfer properties of ammonium nitrate solution in a cation exchange membrane system, providing insights into its applications in electrochemical processes and membrane technology (Kozaderova O et al., 2022).

Analysis Note

Assay: 0.99 - 1.01 mol/l
pH-value: 5.0 - 6.0
As (Arsenic): ≤ 0.005 ppm
Be (Beryllium): ≤ 0.001 ppm
Cd (Cadmium): ≤ 0.002 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.005 ppm
Cu (Copper): ≤ 0.005 ppm
Hg (Mercury): ≤ 0.001 ppm
Mg (Magnesium): ≤ 0.001 ppm
Mo (Molybdenum): ≤ 0.005 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.005 ppm
Sb (Antimony): ≤ 0.005 ppm
Tl (Thallium): ≤ 0.005 ppm
U (Uranium): ≤ 0.001 ppm
V (Vanadium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.005 ppm

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Liq. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service