Skip to Content
Merck
All Photos(1)

Documents

1.02307

Sigma-Aldrich

α-Chymotrypsin from bovine pancreas

cryst. lyophilized 350 U/mg for biochemistry

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21

form

solid

potency

>4000 mg/kg LD50, oral (rat)

pH

3-5 (10 g/L in H2O)

storage temp.

2-8°C

General description

α-Chymotrypsin (CT) is a member of the S1 family. It comprises a catalytic triad system including aspartate, histidine, and serine. It is produced by chymotrypsinogen in the pancreas and then secreted into the digestive tract.

Application

α-Chymotrypsin has been used to study its adsorption characteristics on solid/liquid interfaces under varying adsorption conditions.

Biochem/physiol Actions

α-Chymotrypsin (CT) closely denotes a group of digestive enzymes (trypsin, elastase) that hydrolyzes polypeptide chains. The catalytic triad system works together to regulate the nucleophilicity of the serine residue in catalysis. CT shows a capability to catalyze nonconventional synthetic reactions in water-miscible organic media. Additionally, it is also used as a tool in comparative studies of water-miscible and water-immiscible solvents.

Features and Benefits

  • Easily obtainable due to small size
  • Relatively stable

Analysis Note

Appearance (description):lyophilisate
Appearance (colour):slightly yellowish
Activity (ATEE, pH 8.0, 25 °C):≥ 350 U/mg
Spec. activity (calc. on protein):ca. 400

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service