Skip to Content
Merck
All Photos(1)

Key Documents

1.00826

Supelco

Boron Test, photometric

photometric, 0.05-2.00 mg/L (B), Spectroquant®

Synonym(s):

Trihydroxidoboron, Orthoboric acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
CHF 253.00

CHF 253.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
CHF 253.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

CHF 253.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest more information

Product Name

Boron Cell Test, photometric, 0.05-2.00 mg/L (B), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

boron

measuring range

0.05-2.00 mg/L (B)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Azomethine H)

storage temp.

15-25°C

General description

Method: photometric 0.05 - 2.00 mg/l B Spectroquant®

The method is analogous to DIN 38405-17.This Spectroquant® Boron Cell Test allows the accurate quantification of the Boron content in aqueous samples. Method applied: In weakly acidic solutions borate reacts with azomethine H to form a yellow compound that is determined photometrically. The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided. All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service