07-376
Anti-acetyl-Histone H2A Antibody
serum, Upstate®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Specificity
acetylated Histone H2A
Immunogen
peptide containing surrounding acetylated lysine residues 5, 9, 13 and 15 of human Histone H2A.
Application
Detect acetyl-Histone H2A also known as Histone H2A with Anti-acetyl-Histone H2A Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in WB.
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Histones
Histones
Quality
routinely evaluated by immunoblot acid extracted proteins from sodium butyrate treated HeLa cells (Catalog # 13-113)
Target description
14 kDa
Physical form
0.05% sodium azide and 30% glycerol
Antiserum
Storage and Stability
2 years at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Optics express, 29(20), 32691-32699 (2021-10-08)
We demonstrate single-shot nondiffracting light-sheet microscopy by the incoherent superposition of dispersed polychromatic light sources. We characterized our technique by generating a Bessel light-sheet with a supercontinuum light-source and a C-light-sheet using a diode laser, and demonstrated its applicability to
Journal of nanobiotechnology, 22(1), 223-223 (2024-05-04)
Cardiac muscle targeting is a notoriously difficult task. Although various nanoparticle (NP) and adeno-associated viral (AAV) strategies with heart tissue tropism have been developed, their performance remains suboptimal. Significant off-target accumulation of i.v.-delivered pharmacotherapies has thwarted development of disease-modifying cardiac
Molecular biology of the cell, 35(1), ar12-ar12 (2023-11-22)
Chromosome segregation relies on the correct assembly of a bipolar spindle. Spindle pole self-organization requires dynein-dependent microtubule (MT) transport along other MTs. However, during M-phase RanGTP triggers MT nucleation and branching generating polarized arrays with nonastral organization in which MT
Nature cell biology, 25(10), 1453-1464 (2023-09-29)
Integrin-mediated focal adhesions are the primary architectures that transmit forces between the extracellular matrix (ECM) and the actin cytoskeleton. Although focal adhesions are abundant on rigid and flat substrates that support high mechanical tensions, they are sparse in soft three-dimensional
The specification and global reprogramming of histone epigenetic marks during gamete formation and early embryo development in C. elegans.
PLoS Genetics null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service