Skip to Content
Merck
All Photos(1)

Key Documents

07-2276-I

Sigma-Aldrich

Anti-mSin1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1, Mitogen-activated protein kinase 2-associated protein 1, Stress-activated map kinase-interacting protein 1, SAPK-interacting protein 1, mSIN1, mSin1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), feline (based on 100% sequence homology), horse (based on 100% sequence homology), hamster (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAPKAP1(79109)

General description

Target of rapamycin complex 2 subunit MAPKAP1 (UniProt Q9BPZ7; also known as MEKK2-interacting protein 1, Mitogen-activated protein kinase 2-associated protein 1, mSIN1, Ras inhibitor MGC2745, SAPK-interacting protein 1, Stress-activated protein kinase-interacting 1, Stress-activated map kinase-interacting protein 1,, TORC2 subunit MAPKAP1) is encoded by the MAPKAP1 (also known as JC310, MIP1 SIN1, SIN1b, SIN1g) gene (Gene ID 79109) in human. mSIN1 is an essential component of the Torc2 complex. mTORC2 is comprised of mTOR, a large Ser/Thr protein kinase, along with mSIN1, GbL (mLST8), Protor1, Protor2, and Rictor. The mTORC2 complex is activated primarily downstream of PI3 Kinase and is known to affect cell proliferation and survival primarily by phosphorylating Akt on Ser473. Additionally, the mTORC2 complex is known to affect cytoskeletal organization and cell migration by exerting its effects through Rac, Rho, and PKC. mSIN1 is also known to directly interact with Ras, SPAKs, and JNK.

Specificity

Expected to react with spliced isoforms 1, 2, 3, 5, 6, but not 4.

Immunogen

Epitope: MAP3K2/NBN interaction domain
KLH-conjugated linear peptide corresponding to the MAP3K2/NBN interaction domain of human mSin1.

Application

Detect MAPKAP1/Sin1 using this rabbit polyclonal Anti-mSin1 Antibody, Cat. No. 07-2276-I, validated for use in Western Blotting and Immunoprecipitation.
Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated mSin1 in 0.5 mg of A431 cell lysate.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected mSin1 in 10 µg of A431 cell lysate.

Target description

~59 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasir H Ibrahim et al.
The Journal of biological chemistry, 299(5), 104644-104644 (2023-03-26)
The mechanistic target of rapamycin complex 1 (mTORC1) is a central regulator of mammalian cell growth that is dysregulated in a number of human diseases, including metabolic syndromes, aging, and cancer. Structural, biochemical, and pharmacological studies that have increased our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service