Skip to Content
Merck
All Photos(2)

Documents

05-808

Sigma-Aldrich

Anti-dimethyl-Histone H3 (Arg2) Antibody, rabbit monoclonal

Upstate®, from rabbit

Synonym(s):

H3R2me2, Histone H3 (di methyl R2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

dot blot: suitable
inhibition assay: suitable (peptide)
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Arg2)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.



The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

peptide containing the sequence AdimRTKQ, corresponding to dimethyl-arginine 2 of human histone H3.

Application

Use Anti-dimethyl-Histone H3 (Arg2) Antibody (Rabbit Monoclonal Antibody) validated in DB, WB, PIA, Mplex to detect dimethyl-Histone H3 (Arg2) also known as H3R2me2, Histone H3 (di methyl R2).

Quality

routinely evaluated in acid-extracted proteins from HeLa cells

Target description

17kDa

Physical form

Format: Purified

Analysis Note

Control
Acid extracts from HeLa cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zichuan Liu et al.
Developmental biology, 368(2), 304-311 (2012-06-05)
Several research groups have suggested that the embryonic-abembryonic (Em-Ab) axis in the mouse can be predicted by the first cleavage plane of the early embryo. Currently, it is not known whether this early patterning occurs in cloned embryos produced by
Lessly P Sepulveda-Rincon et al.
Biology of reproduction, 95(6), 123-123 (2016-10-21)
The first lineage specification during mammalian embryo development can be visually distinguished at the blastocyst stage. Two cell lineages are observed on the embryonic-abembryonic axis of the blastocyst: the inner cell mass and the trophectoderm. The timing and mechanisms driving
Sitaram Gayatri et al.
Scientific reports, 6, 28718-28718 (2016-06-25)
Signal transduction in response to stimuli relies on the generation of cascades of posttranslational modifications that promote protein-protein interactions and facilitate the assembly of distinct signaling complexes. Arginine methylation is one such modification, which is catalyzed by a family of
Alessandra Di Lorenzo et al.
Nucleic acids research, 42(13), 8297-8309 (2014-06-19)
Protein arginine methyltransferase 6 (PRMT6) is a nuclear enzyme that modifies histone tails. To help elucidate the biological function of PRMT6 in vivo, we generated transgenic mice that ubiquitously express PRMT6 fused to the hormone-binding portion of the estrogen receptor
Ayaz Ahmad et al.
PloS one, 6(8), e22664-e22664 (2011-08-20)
Post-translational methylation of arginine residues profoundly affects the structure and functions of protein and, hence, implicated in a myriad of essential cellular processes such as signal transduction, mRNA splicing and transcriptional regulation. Protein arginine methyltransferases (PRMTs), the enzymes catalyzing arginine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service